Learn More
Cathepsin D Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06114
Catalog No. 06-114-MI
Add to cart
Description
Description
Anti-Cathepsin D Antibody is an antibody against Cathepsin D for use in western blotting and immunocytochemistry.
Specifications
Specifications
Cathepsin D
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human
Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
CTSD; CLN10; CPSD
GST-tagged recombinant protein corresponding to human Cathepsin D at the heavy chain.
100μg
Primary
NP_001900