Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathespin D Mouse anti-Human, Unlabeled, Clone: 49, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610800

 View more versions of this product

Catalog No. BDB610800


Add to cart

Description

Description

Cathepsin D, an enzyme that degrades proteins, was originally cloned during the search of estrogen responsive genes in MCF-7 cells. Cathepsin D is synthesized as the 43kDa preprocathepsin D that is cleaved to form a 46kDa glycosylated procathepsin D. Procathepsin is then processed into a 44kDa active Cathepsin D. The active and mature forms undergo a further cleavage that yields 28kDa and 15kDa (heavy and light chains, respectively) fragments in SDS-PAGE. The heavy and light chains of Cathepsin D are released into the extracellular medium. The maturation process of Cathepsin D occurs through the transit from the endoplasmic reticulum, Golgi apparatus, and to the lysosomes. Estrogens stimulate cell proliferation in a number of tumor cell lines and anti-estrogen therapy is often used in the treatment of breast cancer patients. Therefore, Cathepsin D, which is estrogen inducible, may have a role during the pathogenesis of breast tumors. Additionally, several other roles have been proposed for this enzyme, such as tissue remodeling, tumor invasion, and embryo implantation.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Cathespin D
49
Unconjugated
Affinity Purified
Human Cathespin D
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.