Learn More
Caveolin-1 (pY14) Mouse, Unlabeled, Clone: 56, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 611339
Description
Caveolin (VIP21) localizes to non-clathrin membrane invaginations (caveolae) on the inner surface of the plasma membrane. In addition, it is present in the trans-Golgi network (TGN) and in apically and basolaterally destined transport vesicles. Caveolin is a transmembrane adaptor molecule that recognizes GPI-linked proteins and interacts with downstream cytoplasmic signaling molecules, such as src-family tyrosine kinases and hetero-trimeric G proteins. Caveolin forms large lipid-binding oligomers, which are thought to play a role in caveolae formation. It may also function as a scaffolding protein, which organizes signaling molecules. This functional role is supported by the fact that caveolin interacts directly with inactive ras and G-protein α subunits. Phosphorylation of caveolin at Tyr-14, Ser-88, and other residues in v-src transformed cells leads to flattening, aggregation, and fusion of caveolae and caveolae-derived vesicles. Thus, caveolin is the principle protein of caveolae and may be involved in v-src mediated cellular transformation.
Host Species: Mouse
Clone: 56
Isotype: IgG1
Species Reactivity: Human
Immunogen: Phosphorylated Human Caveolin-1 (Y14) Peptide
Formula Weight [Chemical]: 21-24kDa
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Intracellular Staining, Western Blotting
Specifications
Caveolin-1 (pY14) | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Phosphorylated Human Caveolin-1 (Y14) Peptide | |
150 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
56 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.