Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-1 (pY14) Mouse, Unlabeled, Clone: 56, BD

Mouse Monoclonal Antibody

$310.00 - $530.00

Specifications

Antigen Caveolin-1 (pY14)
Isotype IgG1
Immunogen Phosphorylated Human Caveolin-1 (Y14) Peptide
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB611338
View Documents
BD Biosciences
611338
50μg
Each for $310.00
Add to cart
 
BDB611339
View Documents
BD Biosciences
611339
150μg
Each for $530.00
Add to cart
 
Description

Description

Caveolin (VIP21) localizes to non-clathrin membrane invaginations (caveolae) on the inner surface of the plasma membrane. In addition, it is present in the trans-Golgi network (TGN) and in apically and basolaterally destined transport vesicles. Caveolin is a transmembrane adaptor molecule that recognizes GPI-linked proteins and interacts with downstream cytoplasmic signaling molecules, such as src-family tyrosine kinases and hetero-trimeric G proteins. Caveolin forms large lipid-binding oligomers, which are thought to play a role in caveolae formation. It may also function as a scaffolding protein, which organizes signaling molecules. This functional role is supported by the fact that caveolin interacts directly with inactive ras and G-protein α subunits. Phosphorylation of caveolin at Tyr-14, Ser-88, and other residues in v-src transformed cells leads to flattening, aggregation, and fusion of caveolae and caveolae-derived vesicles. Thus, caveolin is the principle protein of caveolae and may be involved in v-src mediated cellular transformation.

Host Species: Mouse
Clone: 56
Isotype: IgG1
Species Reactivity: Human
Immunogen: Phosphorylated Human Caveolin-1 (Y14) Peptide
Formula Weight [Chemical]: 21-24kDa

Flow Cytometry, Immunofluorescence, Immunohistochemistry, Intracellular Staining, Western Blotting

Specifications

Specifications

Caveolin-1 (pY14)
Phosphorylated Human Caveolin-1 (Y14) Peptide
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
56
250μg/mL
SDS

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit