Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf18 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524004

Catalog No. PIPA524004

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The exact function of C2orf18 remains unknown. It contains 1 DUF6 domain.
Specifications

Specifications

C2orf18
Polyclonal
PBS with 0.09% sodium azide
Q8N357
ANT2BP, ANT2-binding protein, CB018
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54978
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SLC35F6
Liquid
SLC35F6
KLH conjugated synthetic peptide between 329-358 amino acids from the C-terminal region of human CB018
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit