Learn More
C/EBP zeta, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA124332
Catalog No. PIPA124332
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Stimulates transcription from the HSP70 promoter.
PA1-24332 detects CBF in human samples.PA1-24332 has been successfully used in Western Blot procedures.PA1-24332 immunogen is a synthetic peptide region of human CEBPZ.
Specifications
Specifications
CBF
Polyclonal
Unconjugated
CEBPZ
Purified
Rabbit
IgG
100μg
Avoid Freeze/Thaw Cycles
Primary
10153
Western Blot
1mg/mL
PBS with 2% sucrose and no preservative
Q03701
CEBPZ
Synthetic peptide to a region of human CEBPZ
Protein A/G
RUO
Antibody
Polyclonal
Human