Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBL, Mouse, Clone: 6D12, Abnova™

Mouse monoclonal antibody raised against a partial recombinant CBL.

Manufacturer:  Abnova Corporation H00000867M01A

Catalog No. 89-016-463


  Check Availability
Add to cart

Specifications

Description

The cbl oncogene was first identified as part of a transforming retrovirus which induces mouse pre-B and pro-B cell lymphomas. As an adaptor protein for receptor protein-tyrosine kinases, it positively regulates receptor protein-tyrosine kinase ubiquitination in a manner dependent upon its variant SH2 and RING finger domains. Ubiquitination of receptor protein-tyrosine kinases terminates signaling by marking active receptors for degradation. [provided by RefSeq

Sequence: NIQSQAPSITESSTFGEGNLAAAHANTGPEESENEDDGYDVPKPPVPAVLARRTLSDISNASSSFGWLSLDGDPTTNVTEGSQVPERPPKPFPRRINSER
Description & Specifications

Specifications

CBL
6D12
Cas-Br-M (murine) ecotropic retroviral transforming sequence
CBL
Liquid
CBL
CBL (NP_005179, 736 a.a. ∽836 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26kDa.
200μL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA,Immunoprecipitation,Western Blot
Unlabeled
In ascites fluid
NM_005188
C-CBL, CBL2, RNF55
Murine
IgG1, κ
RUO
Primary
867
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit