Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522273

Catalog No. PIPA522273


Add to cart

Description

Description

PA5-22273 targets CBS in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-22273 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 39 and 251 (P35520) of CBS. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

CBS
Polyclonal
Unconjugated
CBS
Liquid
CBS
Recombit fragment corresponding to a region within amino acids 39 and 251 of CBS (Uniprot IDNo.P35520)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
P35520
HIP4, beta-thionase, cystathionine beta-synthase, methylcysteine synthase, serine sulfhydrase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
875
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit