Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBX2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526611
Description
This antibody is predicted to react with mouse based on sequence homology.
CBX2 is a component of the polycomb multiprotein complex, which is required to maintain the transcriptionally repressive state of many genes throughout development via chromatin remodeling and modification of histones.Specifications
CBX2 | |
Polyclonal | |
CBX2 | |
CDCA6, M33, SRXY5, Pc class homolog, cell division cycle associated 6, chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, modifier 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84733 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q14781 | |
CBX2 | |
KLH conjugated synthetic peptide between 47-75 amino acids from the N-terminal region of human CBX2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction