Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chromobox homolog 4 (Pc class homolog, Drosophila), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CBX4 protein.

Manufacturer:  Abnova Corporation H00008535B01

Catalog No. 89-006-580


Add to cart

Description

Description

Sequence: MELPAVGEHVFAVESIEKKRIRKGRVEYLVKWRGWSPKYNTWEPEENILDPRLLIAFQNRERQEQLMGYRKRGPKPKPLVVQVPTFARRSNVLTGLQDSSTDNRAKLDLGAQGKGQGHQYELNSKKHHHHAVGLNLSHVRKRCLSETHGEREPCKKRLTARSISTPTCLGGSPAAERPADLPPAAALPQPEVILLDSDLDEPIDLRCVKTRSEAGEPPSSLQVKPETPASAAVAVAAAAAPTTTAEKPPAEAQDEPAESLSEFKPFFGNIIITDVTANCLTVTFKEYVTV
Specifications

Specifications

chromobox homolog 4 (Pc class homolog, Drosophila)
Unconjugated
No additive
BC014967.1
NBP16|PC2|hPC2
Mouse
50 uL
Apoptosis
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human CBX4 protein.
CBX4
Serum
CBX4
CBX4 (AAH14967.1, 1 a.a. ~ 290 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8535
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit