Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP1 alpha Rabbit anti-Canine, Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523357

Catalog No. PIPA523357


Add to cart

Description

Description

CBX5, also known as Chromobox protein homolog 5, is a regulatory protein belonging to the Polycomb group proteins family with two chromo domains. CBX5 associates with chromosomes during mitosis, specifically with chromatin during metaphase and anaphase. A component of the centromeric and pericentromeric heterochromatin, CBX5 may interact with lamin B receptor (LBR) to contribute to the association of the heterochromatin with the inner nuclear membrane. CBX5 recognizes and binds histone H3 tails methylated at  ft.Lys-9 ft., leading to epigenetic repression. This methyl-lysine binding protein localized at heterochromatin sites is also known to mediate gene silencing and is involved in the formation of functional kinetochore through interaction with MIS12 complex proteins.
Specifications

Specifications

HP1 alpha
Polyclonal
Unconjugated
CBX5
Liquid
CBX5
Synthetic peptide (176 AYPEDAENKEKETAKS 191) of CBX5 protein.
Protein A
RUO
Antibody
Polyclonal
Canine, Human, Mouse, Non-human primate
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P45973, Q61686
HP1, HP1A, HP1 alpha homolog, HP1-ALPHA, HP1Hs alpha, antigen p25, chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12419, 23468, 477593
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit