Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CBX6, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128731

Catalog No. NBP128731


Add to cart

Description

Description

CBX6 Polyclonal antibody specifically detects CBX6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

CBX6
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry 1:500-1:2000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500-1:2000
NP_055107.3
chromobox homolog 6, chromobox protein homolog 6
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
CBX6
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human chromobox homolog 6 using the numbering given in entry NP_055107.3 (GeneID 23466).
Immunogen affinity purified
RUO
Primary
23466
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only