Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIP30 Rabbit anti-Canine, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124473

Catalog No. PIPA124473


Add to cart

Description

Description

PA1-24473 detects CC3 (HTATIP2) in Human samples. PA1-24473 has been successfully used in Western Blot and immunohistochemistry (paraffin) procedures. While diluting for Western blot, use a 5% skim milk/PBS buffer solution. PA1-24473 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human HTATIP2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

CC3 (HTATIP2) is a member of the short-chain dehydrogenases/reductases (SDR) family. It is a novel serine/threonine kinase that phosphorylates the C-terminal domain (CTD) of the largest RNA polymerase II subunit and induces the expression of apoptosis related genes Bad and Siva, as well as metastasis suppressor NM23-H2. It also interacts with an estrogen receptor alpha-interacting coactivator CIA and regulates ERalpha-mediated c-myc transcription.
Specifications

Specifications

TIP30
Polyclonal
Unconjugated
HTATIP2
Liquid
HTATIP2
Synthetic peptide region of human HTATIP2.
IgG fraction
RUO
Antibody
Polyclonal
Canine, Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9BUP3, Q9Z2G9
HIV-1 tat interactive protein 2, CC3
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10553, 476886, 53415
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit