Learn More
TMEM30B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524890
Catalog No. PIPA524890
Add to cart
Specifications
Specifications
TMEM30B
Polyclonal
Unconjugated
TMEM30B
Liquid
TMEM30B
KLH conjugated synthetic peptide between 42-71 amino acids from the N-terminal region of human CC50B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q3MIR4
CDC50B, cell cycle control protein 50B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
161291