Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCD92 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524939

Catalog No. PIPA524939

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CCD92
Polyclonal
PBS with 0.09% sodium azide
Q53HC0, Q8VDN4
coiled-coil domain-containing protein 92, limkain beta 2, limkain beta-2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
215707, 80212
Immunohistochemistry, Western Blot
Unconjugated
CCDC92
Liquid
CCDC92
KLH conjugated synthetic peptide between 177-205 amino acids from the central region of human CCD92
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit