Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC126 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524334

Catalog No. PIPA524334

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Specifications

Specifications

CCDC126
Polyclonal
PBS with 0.09% sodium azide
Q96EE4, Q8BIS8
coiled-coil domain-containing protein 126
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57895, 90693
Western Blot
Unconjugated
CCDC126
Liquid
CCDC126
KLH conjugated synthetic peptide between 77-106 amino acids from the C-terminal region of human CCDC126
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit