Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC146 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525376

Catalog No. PIPA525376

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function ofCCDC146 remains unknown.
Specifications

Specifications

CCDC146
Polyclonal
PBS with 0.09% sodium azide
Q8IYE0
coiled-coil domain-containing protein 146
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57639
Western Blot
Unconjugated
CCDC146
Liquid
CCDC146
KLH conjugated synthetic peptide between 113-141 amino acids from the N-terminal region of human CCDC146
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit