Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC148 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524107

Catalog No. PIPA524107


Add to cart

Description

Description

There are three different isoforms.
Specifications

Specifications

CCDC148
Polyclonal
PBS with 0.09% sodium azide
Q8NFR7, Q6P5U8
coiled-coil domain-containing protein 148
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
130940, 227933
Flow Cytometry, Western Blot
Unconjugated
CCDC148
Liquid
CCDC148
KLH conjugated synthetic peptide between 279-307 amino acids from the central region of human CCDC148
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit