Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC38 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524141
Description
This antibody is predicted to react with non-human primate based on sequence homology.
CCDC38 Polyclonal antibody specifically detects CCDC38 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CCDC38 | |
Polyclonal | |
Unconjugated | |
Ccdc38 | |
4933417K05Rik; CCDC38; coiled-coil domain containing 38; coiled-coil domain-containing protein 38; RGD1564046 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
120935, 237465 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q502W7, Q8CDN8 | |
Ccdc38 | |
KLH conjugated synthetic peptide between 352-380 amino acids from the central region of human CCDC38 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction