Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC39 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524516

Catalog No. PIPA524516

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CCDC39
Polyclonal
PBS with 0.09% sodium azide
Q9UFE4, Q9D5Y1
coiled-coil domain-containing protein 39
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100750499, 339829, 51938
Western Blot
Unconjugated
CCDC39
Liquid
CCDC39
KLH conjugated synthetic peptide between 770-798 amino acids from the C-terminal region of human CCDC39
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit