Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

coiled-coil domain containing 6 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant CCDC6.

Manufacturer:  Abnova Corporation H00008030A01

Catalog No. 89-004-377


Add to cart

Description

Description

Sequence: HTVGFTPPTSLTRAGMSYYNSPGLHVQHMGTSHGITRPSPRRSNSPDKFKRPTPPPSPNTQTPVQPPPPPPPPPMQPTVPSAATSQPTPSQHSAHPSSQP
Specifications

Specifications

coiled-coil domain containing 6
Unconjugated
50% glycerol
NM_005436
D10S170|FLJ32286|H4|PTC|TPC|TST1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8030
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant CCDC6.
CCDC6
Serum
CCDC6
CCDC6 (NP_005427, 375 a.a. ~ 474 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit