Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC69 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525380

Catalog No. PIPA525380

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of CCDC69 remains unknown.
Specifications

Specifications

CCDC69
Polyclonal
PBS with 0.09% sodium azide
A6NI79, Q3TCJ8
coiled-coil domain-containing protein 69
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26112, 52570
Flow Cytometry, Western Blot
Unconjugated
CCDC69
Liquid
CCDC69
KLH conjugated synthetic peptide between 141-170 amino acids from the central region of human CCDC69
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit