Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCDC82, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128743

Catalog No. NBP128743


Add to cart

Description

Description

CCDC82 Polyclonal antibody specifically detects CCDC82 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

CCDC82
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 10 ug/mg lysate
NP_079001.2
coiled-coil domain containing 82, coiled-coil domain-containing protein 82, FLJ23518, HSPC048
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
CCDC82
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human coiled-coil domain containing 82 using the numbering given in entry NP_079001.2 (GeneID 79780).
Immunogen affinity purified
RUO
Primary
79780
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only