Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCKAR Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124544

Catalog No. PIPA124544


Add to cart

Description

Description

PA1-24544 detects Cholecystokinin A Receptor in Human samples. PA1-24544 has been successfully used in Immunohistochemistry (paraffin) procedures. PA1-24544 immunogen corresponds to Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 35/55%. Rat: 65/70%.

Cholecystokinin A receptor, a Cholecystokinin Receptor, binds sulfated CCK, particularly CCK8. The CCKA receptor mediates gallbladder contraction, gastric emptying, relaxation of the sphincter of Oddi, and insulin secretion. It is also inolved in pancreatic growth, enzyme secretion, and tumorigenesis. Cholecystokinin A receptor has been reported in brain, colon, gall bladder, gastrointestinal tract, kidney, pancreas, small intestine, spinal cord, and stomach. ESTs have been isolated from mouse kidney and heart libraries.
Specifications

Specifications

CCKAR
Polyclonal
Unconjugated
CCKAR
Liquid
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
886
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
P32238
CCKAR
Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 35/55%. Rat: 65/70%.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit