Learn More
CCKAR Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136144
Catalog No. 01-672-100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A suggested positive control for IHC is rat pancreas. Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.
This gene encodes a G-protein coupled receptor that binds non-sulfated members of the cholecystokinin (CCK) family of peptide hormones. This receptor is a major physiologic mediator of pancreatic enzyme secretion and smooth muscle contraction of the gallbladder and stomach. In the central and peripheral nervous system this receptor regulates satiety and the release of beta-endorphin and dopamine.
Specifications
Specifications
CCKAR
Polyclonal
whole serum with 0.1% sodium azide
O08786, P30551
CCK-A, CCK1-R, CCKRA
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry (Frozen)
Unconjugated
Cckar
Lyophilized
CCKAR
Synthetic peptide from the C-terminal of the rat CCK-A receptor.
50 μL
4° C
Primary
12425, 24889