Learn More
CCKBR, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA131120
Catalog No. PIPA131120
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene encodes a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors.
PA1-31120 detects CCKBR from human samples.PA1-31120 has been successfully used in immunohistochemistry (paraffin tissue), Western blot applications.The PA1-31120 immunogen is: Poly Lysin conjugated synthetic peptide: VHRWPSARVRQTWS, corresponding to amino acids 206-219 of Human CCKBR.
Specifications
Specifications
CCK-B Receptor
Polyclonal
whole serum with no preservative
P32239
CCK-B, CCK2R, GASR
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CCKBR
Serum
CCKBR
Poly Lysin conjugated synthetic peptide: VHRWPSARVRQTWS, corresponding to amino acids 206-219 of Human CCKBR.
50μL
Store at -20°C
Primary
887