Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCL17/TARC, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP140991

Catalog No. NBP140991



Description

Description

CCL17/TARC Polyclonal antibody specifically detects CCL17/TARC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

CCL17/TARC
1.0 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Functional
Q92583
ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine
Rabbit
IgG
0.05 mg
Chemokines and Cytokines
Primary
6361
Human
Western Blot, Flow Cytometry, ELISA, Functional Assay
Unconjugated
PBS (pH 7.2) with 0.09% Sodium Azide
Purified
CCL17
Recombinant human CCL17
IgG purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This antibody specifically recognises human CCL17, otherwise known as TARC (thymus and activation-regulated chemokine), a member of the intercrine beta (chemokine CC) family, predominantly produced by thymic dendritic cells, which specifically acts as a chemotactic factor for T cells, and signals through binding to the CCR4 receptor.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only