Learn More
MCP-1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA140299
Catalog No. 01-676-418
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This chemotactic factor attracts monocytes and basophils but not neutrophils or eosinophils and augments monocyte anti-tumor activity. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis or atherosclerosis. It may be involved in the recruitment of monocytes into the arterial wall during the disease process of atherosclerosis.
PA1-40299 detects MCP-1 from human samples.PA1-40299 has been successfully used in ELISA, functional assays, immunohistochemistry (frozen), immunoprecipitation, and Western blot applications.The PA1-40299 imunogen is human MCP-1.
Specifications
Specifications
MCP-1
Polyclonal
Unconjugated
CCL2
Liquid
CCL2
Human MCP-1.
Purified
RUO
Antibody
Polyclonal
Human
ELISA, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P13500
GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, MGC9434, SCYA2, SMC-CF
Rabbit
IgG
100 μg
4° C
Primary
6347