Learn More
MCP-1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA135854
Catalog No. 01-672-355
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
Description
Description
MCP-1 is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
Specifications
Specifications
MCP-1
Polyclonal
Unconjugated
CCL2
Liquid
CCL2
Human MCP-1.
Purified
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P13500
CCL2
Rabbit
IgG
100 μg
4° C
Primary
6347