Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL20, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against full-length recombinant CCL20.

Manufacturer:  Abnova Corporation PAB16925

Catalog No. 89-024-836


Add to cart

Specifications

Specifications

CCL20
Unconjugated
The optimal working dilution should be determined by the end user.
CCL20
CKb4|LARC|MIP-3a|MIP3A|SCYA20|ST38
Rabbit
100 ug
Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water
Polyclonal
Human
ELISA, Western Blot
Rabbit polyclonal antibody raised against full length recombinant CCL20.
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2
Lyophilized
CCL20
Recombinant protein corresponding to full length human CCL20.
RUO
Primary
6364
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit