Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCL20/MIP-3 alpha, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142763

Catalog No. NBP142763


Add to cart

Description

Description

CCL20/MIP-3 alpha Polyclonal antibody specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Western Blot, ELISA, Block/Neutralize.
Specifications

Specifications

CCL20/MIP-3 alpha
LYOPH
Western Blot 1:500-1:2000, ELISA 1:20000-1:100000, Blocking/Neutralizing 50-200 molar excess
P78556
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38
Rabbit
IgG
0.1 mg
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
In ELISA and other immunoreactive assays, this antibody will recognize both native and recombinant human MIP-3a in cell supernatants and certain body fluids. A control of similarly diluted normal rabbit IgG is recommended.
Human
Blocking Assay, ELISA, Western Blot
Unconjugated
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
Purified
CCL20
This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human MIP-3 alpha protein.
IgG purified
RUO
Primary
6364
Reconstitute with 100 ul deionized water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only