Learn More
CCL3 Goat anti-Mouse, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA124657
Catalog No. PA124657
Add to cart
Description
Description
PA1-24657 detects Macrophage Inflammatory Protein 1 alpha from mouse samples. PA1-24657 has been successfully used in neutralization, inhibition assays, ELISA, and Western blot procedures. Mouse MIP1 alpha was tested for its ability to neutralize the bioactivity of recombit, mouse MIP1 alpha in a cell proliferation assay using hematopoietic stem cells. The ND50 of the antibody is defined as the concentration of antibody resulting in a one-half maximal inhibition of bioactivity of recombit, mouse MIP1 alpha, which is present at five times its own EC50 (the concentration of recombit, mouse MIP1 alpha producing a one-half maximal bioactivity without antibody). The PA1-24657 immunogen is recombit mouse macrophage inflammatory protein-1alpha, expressed in E. coli.
Specifications
Specifications
CCL3
Polyclonal
Unconjugated
Ccl3
Liquid
CCL3
Recombit mouse macrophage inflammatory protein-1alpha, expressed in E. coli.
Protein G
RUO
Antibody
Polyclonal
Mouse
ELISA, Inhibition Assays, Neutralization, Western Blot
1 mg/mL
PBS with no preservative; pH 7.4
P10855
RP23-320E6.7, AI323804, G0S19-1, LD78alpha, MIP-1alpha, MIP1-(a), MIP1-alpha, Mip1a, Scya3
Goat
IgG
100 μg
4° C, do not freeze
Primary
20302