Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128899

Catalog No. PIPA128899


Add to cart

Description

Description

PA1-28899 detects CCL3 from mouse samples. PA1-28899 has been successfully used in Western blot applications. The PA1-28899 immunogen is: Recombit mouse MIP-1 alpha

Specifications

Specifications

CCL3
Polyclonal
Unconjugated
Ccl3
Lyophilized
CCL3
Recombit mouse MIP-1 alpha
Affinity chromatography
RUO
Antibody
Polyclonal
Mouse
ELISA, Western Blot
1 mg/mL
PBS with no preservative; pH 7.4
P10855
RP23-320E6.7, AI323804, G0S19-1, LD78alpha, MIP-1alpha, MIP1-(a), MIP1-alpha, Mip1a, Scya3
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20302
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit