Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129045

Catalog No. PIPA129045


Add to cart

Description

Description

PA1-29045 detects CCL3 from human samples. PA1-29045 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-29045 immunogen is: Synthetic peptide: EEW VQKYVSDLEL SA , corresponding to C terminal amino acids 77-92 of Human Macrophage Inflammatory Protein 1 alpha / CCL3

Macrophage inflammatory protein-1 is a so-called monokine that is involved in the acute inflammatory state in the recruitment and activation of polymorphonuclear leukocytes (Wolpe et al., 1988 [PubMed 3279154]). Sherry et al. (1988) [PubMed 3058856] demonstrated 2 protein components of MIP1, called by them alpha and beta.
Specifications

Specifications

CCL3
Polyclonal
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
P10147
G0S19-1, LD78ALPHA, MIP-1-alpha, MIP1A, SCYA3
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
6348
Immunohistochemistry (Paraffin)
Unconjugated
CCL3
Liquid
CCL3
synthetic peptide EEW VQKYVSDLEL SA , corresponding to C terminal amino acids 77-92 of Human Macrophage Inflammatory Protein 1 alpha / CCL3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit