Learn More
CCL4L1/CCL4L2 Mouse anti-Human, Clone: 4G8, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA523300
Catalog No. PIMA523300
Add to cart
Description
Description
Peptide Sequence: SDPPTACCFS YTARKLPRNF VVDYYETSSL CSQPAVVFQT KRGKQVCADP SESWVQEYVY DLELN*
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. This protein is similar to CCL4 which inhibits HIV entry by binding to the cellular receptor CCR5. The copy number of this gene varies among individuals; most individuals have 1-5 copies in the diploid genome, although rare individuals do not contain this gene at all. The human genome reference assembly contains two copies of this gene. This record represents the more telomeric gene.
Specifications
Specifications
CCL4L1/CCL4L2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8NHW4
CCL4L1
CCL4L1 (NP_996890, 28 a.a. ∽ 93 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
4G8
Unconjugated
CCL4L1
Liquid
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
388372