Learn More
anti-CCL4L1/LAG-1, HRP, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP177571
Catalog No. NBP177571
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CCL4/MIP-1 beta
HRP
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate
Delipidation and Defibrination
CCL4
This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human MIP1 beta protein.
Delipidation and Defibrination
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
ELISA, Western Blot
Western Blot 1:2000-1:5000, ELISA 1:20000-1:100000
Q8NHW4
ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP-1 beta/CCL4, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2
Rabbit
IgG
0.1 mg
Biologically Active Proteins, Chemokines and Cytokines
Primary
6351