Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL5/RANTES Mouse anti-Human, Clone: 21445, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen CCL5/RANTES
Immunogen E. coli-derived recombinant human CCL5/RANTES
Clone 21445
Applications Western Blot, Immunohistochemistry, Neutralization, Flow Cytometry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB278SP
View Documents
R&D Systems
MAB278SP
25 ug
Each for $125.00
Add to cart
 
MAB278100
View Documents
R&D Systems
MAB278100
100 ug
Each for $377.00
Add to cart
 
MAB278500
View Documents
R&D Systems
MAB278500
500 ug
Each for $839.00
Add to cart
 
Description

Description

CCL5/RANTES Monoclonal antibody MAB278 Specifically detects CCL5/RANTES in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunohistochemistry, Neutralization, Western Blotting
Specifications

Specifications

CCL5/RANTES
21445
Unconjugated
Mouse
IgG1
6352
RUO
E. coli-derived recombinant human CCL5/RANTES
Western Blot, Immunohistochemistry, Neutralization, Flow Cytometry, CyTOF
Purified
Monoclonal
Primary
Detects human CCL5/RANTES in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with other chemokines, including recombinant human CCL2, 3, 4, recombinant mouse (rm) CCL3, and rmCCL4.
Protein A or G purified from ascites
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit