Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYR61, Polyclonal, Invitrogen™

Catalog No. PIPA125681
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA125681 50 μg
1 options

Catalog No. PIPA125681

Supplier: Thermo Scientific PA125681

Chicken Polyclonal Antibody

The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. In contrast to cyclin A1, which is present only in germ cells, this cyclin is expressed in all tissues tested. This cyclin binds and activates CDC2 or CDK2 kinases, and thus promotes both cell cycle G1/S and G2/M transitions.

PA1-25681 detects CCN1 from human, mouse, and rat samples.PA1-25681 has been successfully used in Western blot procedures.The PA1-25681 immunogen is a recombinant fragment corresponding to residues 211-377 of human CCN1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCN1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene CYR61
Gene Accession No. O00622, P18406, Q9ES72
Gene Alias CCN1, CCNA
Gene Symbols CYR61
Host Species Chicken
Immunogen Recombinant fragment corresponding to residues 211-377 of human CCN1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3491
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.