Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR1, FITC, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP178178

Catalog No. NBP178178

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CCR1
FITC
Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% Sodium Azide
Affinity Purified
CCR1
Synthetic peptide taken within amino acid region 1-50 on mouse CCR1 protein.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
The antibody does not cross reacts with other members of the CCR family.
ELISA
ELISA 1:10000
AAH64991
C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
Rabbit
IgG
0.1 mg
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary
1230
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only