Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR1, PE, Clone: 53504, R&D Systems™

Manufacturer:  R&D Systems FAB145P100

 View more versions of this product

Catalog No. FAB145P100


Add to cart

Description

Description

Host Species: Mouse
Clone: 53504
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: Swiss 3T3 mouse cell line transfected with human CCR1.

Applications:

Flow Cytometry

Specifications

Specifications

CCR1
53504
Flow Cytometry 10 uL/10^6 cells
P32246
C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
Mouse
IgG2b
100 Tests
Primary
1230
Human
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR1
Swiss 3T3 mouse cell line transfected with human CCR1, Accession # P32246
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CCR1. Reacts specifically with human CCR1 transfectants and not the parental cell line in flow cytometry. No cross-reactivity with CCR5 transfectants is observed.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit