Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR11, APC, Clone: 674144, R&D Systems™

Manufacturer:  R&D Systems FAB6899A025

 View more versions of this product

Catalog No. FAB6899A025

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Host Species: Mouse
Clone: 674144
Isotype: IgM
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CCR11.

Applications:

Flow Cytometry

Specifications

Specifications

APC
Q9NPB9
25 Tests
Monoclonal
Supplied in a saline solution containing BSA and Sodium Azide.
Affinity Purified
Primary
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit