Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-CCR2, APC, Clone: 48607, R&D Systems™

Manufacturer:  R&D Systems FAB151A025

 View more versions of this product

Catalog No. FAB151A025



Description

Description

Host Species: Mouse
Clone: 48607
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CCR2. Met1-Leu360

Applications:

Flow Cytometry

Specifications

Specifications

CCR2
48607
Flow Cytometry 10 uL/10^6 cells
NP_001116868
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
Mouse
IgG2b
25 Tests
Primary
729230
Human
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR2
NS0 mouse myeloma cell line transfected with human CCR2 Met1-Leu360 Accession # NP_001116868
Protein A or G purified from ascites
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CCR-2 transfectants but not the parental cell line or CCR-5 transfectants.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit