Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR2, DyLight 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP148338V

Catalog No. NBP148338V


Add to cart

Description

Description

CCR2 Polyclonal antibody specifically detects CCR2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CCR2
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
Rabbit
IgG
0.1 ml
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary
729230
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified
CCR2
Synthetic peptide made to an N-terminal portion of the mouse CCR2 protein (within residues 20-100). [Swiss-Prot# P51683]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only