Learn More
CCR2 Rabbit anti-Human, Rat, DyLight® 488, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523040
Catalog No. PIPA523040
Add to cart
Description
Description
CCR2, a Chemokine Receptor, is a regulator of monocyte/macrophage traffickings and is upregulated in response to inflammation. Resistance to HIV-1 infection or delayed progression to AIDS may be linked to CCR2 polymorphisms. The ligand for CCR2 is monocyte chemoattractant protein (MCP-1), a chemokine produced by endothelial cells, smooth muscle cells, and macrophages. Activation of monocytes by MCP-1 may mediate the monocytic infiltration of tissues in atherosclerosis (Boring et al., 1998) and other inflammatory disorders including rheumatoid arthritis, alveolitis, and tumor infiltration. Two isoforms are produced by alternative splicing. CCR2 has been reported to be expressed in a wide variety of tissues, including blood, brain, heart, kidney, liver, lung, ovary, pancreas, spinal cord, spleen, thymus, and vessel. Increased expression of CCR2 has been documented in CD3+ lymphocytes in rheumatoid arthritis (Ruth et al., 2001). ESTs have been isolated from liver/spleen and prostate libraries.
Specifications
Specifications
CCR2
Polyclonal
DyLight™ 488
CCR2
Liquid
CCR2
Synthetic peptide made to an N-terminal portion of the human CCR2 protein (within residues 20-100).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
0.6 mg/mL
sodium borate with 0.05% sodium azide
P41597, O55193
hCG_14621, CC-CKR-2, CCR2A, CCR2B, CD192, CKR2, CKR2A, CKR2B, CMKBR2, MCP-1-R, C-C CKR-2, C-C chemokine receptor type 2, CCR-2, MCP-1 receptor, monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
Rabbit
IgG
100 μL
4° C, store in dark
Primary
60463, 729230