Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR3, Fluorescein, Clone: 61828, R&D Systems™

Manufacturer:  R&D Systems FAB155F025

 View more versions of this product

Catalog No. FAB155F025


Add to cart

Description

Description

Host Species: Rat
Clone: 61828
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: Y3 rat myeloid cell line transfected with human CCR3.

Applications:

Flow Cytometry

Specifications

Specifications

CCR3
61828
Flow Cytometry 10 uL/10^6 cells
P51677
b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841
Rat
IgG2a
25 Tests
Primary
1232
Human
Flow Cytometry
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR3
Y3 rat myeloid cell line transfected with human CCR3 Accession # P51677
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CCR3 transfectants in flow cytometry. In flow cytometry, no cross-reactivity with CCR1, CCR2, CCR5, and CCR6 transfectants or the parental cell line is observed. Detects CCR3 on peripheral blood eosinophils.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit