Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR3 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA121622

Catalog No. PIPA121622


Add to cart

Description

Description

PA1-21622 detects CCR3 in human samples. PA1-21622 has been successfully used in ELISA, immunocytochemistry, immunohistochemistry (paraffin) and Western blot procedures. The PA1-21622 immunogen is a synthetic peptide corresponding to residues C V E T F G T T S Y Y D D V G L L C E K A D T R A L M A of human CCR3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

CCR3 (C-C chemokine receptor type 3) binds to eotaxin, eotaxin-3, MCP-3, MCP-4, RANTES, and MIP-1 delta. CCR3 trasnduces a signal by increasing the intracellular calcium ions level. It also serves as an alternative co-receptor with CD4 for HIV-1 infection. Other chemokines play important roles in inflammation and are critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines.
Specifications

Specifications

CCR3
Polyclonal
Unconjugated
CCR3
Liquid
CCR3
Synthetic peptide corresponding to residues C V E T F G T T S Y Y D D V G L L C E K A D T R A L M A of human CCR3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.1% sodium azide; pH 7.2
P51677
CC-CKR-3, CKR3, CMKBR3, MGC102841
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1232
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit