Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR4 Mouse anti-Human, Clone: KH-4F5-D3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA117770

Catalog No. 01-670-183


Add to cart

Description

Description

This antibody recognizes native human CCR4 protein expressed in transfected mammalian cells.

Chemokines are a group of small (approximately 8 to 14 kD), mostly basic, structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane, G protein-coupled receptors. Chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies, CXC and CC, based on the arrangement of the first 2 of the 4 conserved cysteine residues; the 2 cysteines are separated by a single amino acid in CXC chemokines and are adjacent in CC chemokines.
Specifications

Specifications

CCR4
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.2
P51679
CC-CKR-4, CKR4, CMKBR4, ChemR13, HGCN:14099, K5-5, MGC88293
Mouse
IgG2a
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1233
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
KH-4F5-D3
Unconjugated
CCR4
Liquid
CCR4
Synthetic peptide corresponding to human CCR4.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit