Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR5, Fluorescein, Clone: 45531, R&D Systems™

Manufacturer:  R&D Systems FAB182F100

 View more versions of this product

Catalog No. FAB182F100


Add to cart

Description

Description

Host Species: Mouse
Clone: 45531
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CCR5. Met1-Leu352

Applications:

Flow Cytometry

Specifications

Specifications

CCR5
45531
Flow Cytometry 10 uL/10^6 cells
P51681
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
Mouse
IgG2b
100 Tests
Primary
1234
Human
Flow Cytometry
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR5
NS0 mouse myeloma cell line transfected with human CCR5, Met1-Leu352, Accession # P51681
Protein A or G purified from ascites
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CCR5.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit