Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR5, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP141434

Catalog No. NBP141434

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CCR5
LYOPH
Western Blot 0.1-0.5 ug/ml, Flow Cytometry 1-2 ug/ml, Immunohistochemistry 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml
P51681
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 0.2 ml of distilled water will yield a concentration of 0.5 mg/ml.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3. with 0.05mg Thimerosal
Affinity Purified
CCR5
A synthetic peptide corresponding to a sequence at the N-terminal of human CCR5, different from the related rat sequence by two amino acids.
40 kDa
0.1 mg
Alzheimers Research, Cancer, Chemokine Research, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction
Primary
1234
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only