Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR6, Fluorescein, Clone: 53103, R&D Systems™

Manufacturer:  R&D Systems FAB195F100

 View more versions of this product

Catalog No. FAB195F100


Add to cart

Description

Description

Host Species: Mouse
Clone: 53103
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CCR6. Met1-Met374

Applications:

Flow Cytometry

Specifications

Specifications

CCR6
53103
Flow Cytometry 10 uL/10^6 cells
P51684
BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22
Mouse
IgG2b
100 Tests
Primary
1235
Human
Flow Cytometry
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR6
NS0 mouse myeloma cell line transfected with human CCR6 Met1-Met374 Accession # P51684
Protein A or G purified from ascites
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CCR6. Stains human CCR6 transfectants but not the parental cell line in flow cytometry. Does not cross-react with transfectants expressing human CCR1, CCR2, CCR3, CCR4, CCR5, CCR7, CXCR1, CXCR2, CXCR3, CXCR4, or STRL33.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit